Order Entry
Ireland
ContactUsLinkComponent
Anti-SAD1 (BRSK1) Rabbit Polyclonal Antibody
Anti-SAD1 (BRSK1) Rabbit Polyclonal Antibody
Catalog # ABGEAP8168A
Supplier:  Abgent
undefined
Anti-SAD1 (BRSK1) Rabbit Polyclonal Antibody
Catalog # ABGEAP8168A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BR serine/threonine-protein kinase 1
  • Antigen symbol:
    BRSK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Gene ID:
    645191
  • Antigen synonyms:
    SAD1A|BR serine/threonine-protein kinase 1|SAD1 kinase
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    RERRLQRVTATAGEDC
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    This LINGO3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 395-424 amino acids from the C-terminal region of human LINGO3.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SAD1 (BRSK1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human