Anti-WBP2 Rabbit Polyclonal Antibody
ABGEAP7786A
: Abgent
- Antibody type:Primary
- Antigen name:WW Domain binding Protein 2
- Antigen symbol:WBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:WBP2_HUMAN|WBP 2|MGC18269|WW domain-binding protein 2.|WW domain binding protein 2|Wbp2|WBP-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: WBP2 (WW domain binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human