Anti-SNRK Rabbit Polyclonal Antibody
ABGEAP7249C
: Abgent
- Antibody type:Primary
- Antigen name:SNF related kinase
- Antigen symbol:SNRK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:SNFRK|HSNFRK|SNRK|KIAA0096|FLJ20224|DKFZp779A1866
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SNRK (SNF related kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human