Order Entry
Ireland
ContactUsLinkComponent
 
Anti-DCLK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7219B
Supplier:  Abgent
undefined
Anti-DCLK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7219B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inositol Polyphosphate Multikinase
  • Antigen symbol:
    DCLK1 (doublecortin-like kinase 1)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal (700–800 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    Inositol polyphosphate multikinase|Inositol 1,3,4,6 tetrakisphosphate 5 kinase|IPMK_HUMAN|6-tetrakisphosphate 5-kinase|Ipmk|Inositol 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: DCLK1 (doublecortin-like kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse