Order Entry
Ireland
ContactUsLinkComponent
 
Anti-DONSON Rabbit Polyclonal Antibody
  ABGEAP6567A
 :  Abgent
undefined
Anti-DONSON Rabbit Polyclonal Antibody
  ABGEAP6567A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    K506 binding Protein 1A 12kDa
  • Antigen symbol:
    DONSON
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    PKC12|FKBP-12|FKBP1|PPIASE|PKCI2|FKBP-1A|PPIase FKBP1A|Peptidyl-prolyl cis-trans isomerase FKBP1A|FKBP12|12 kDa FK506-binding protein
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: DONSON (downstream neighbor of SON)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human