Anti-PSG7 Rabbit Polyclonal Antibody
Catalog # ABGEAP4939B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Pregnancy Specific Beta-1-glycoprotein 7
- Antigen symbol:PSG7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:DNA-binding protein TAXREB67|cAMP-dependent transcription factor ATF-4|Cyclic AMP-dependent transcription factor ATF-4|TXREB|TaxREB67|Tax-responsive enhancer element-binding protein 67|CREB2|Activating transcription factor 4|cAMP-responsive element-binding protein 2|CREB-2|Cyclic AMP-responsive element-binding protein 2|ATF4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PSG7 (pregnancy specific beta-1-glycoprotein 7 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human