Anti-CHEK1 Rabbit Polyclonal Antibody
ABGEAP3070A
: Abgent
- Antibody type:Primary
- Antigen name:Checkpoint Kinase 1
- Antigen symbol:CHEK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:kinase Chk1|CHK1|EC 2.7.11.1|CHEK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human