Order Entry
Ireland
ContactUsLinkComponent
Anti-FZD1 Rabbit Polyclonal Antibody
Anti-FZD1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2755C
Supplier:  Abgent
undefined
Anti-FZD1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2755C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Frizzled 1
  • Antigen symbol:
    FZD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    Fz-1|Frizzled-1 [Precursor]|FzE1|hFz1|FZD1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: FZD1 (frizzled class receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human