Anti-UCHL5 Rabbit Polyclonal Antibody
Catalog # ABGEAP2128A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin carboxyl-terminal hydrolase L5
- Antigen symbol:UCHL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-Terminal ( 1–100 aa)
- Cross adsorption:No
- Antigen synonyms:(Ubiquitin C-terminal hydrolase L5|Ubiquitin C-terminal hydrolase|CGI-70|UCH-L5|Ubiquitin thioesterase L5|AD-019|UCH37
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: UCHL5 (ubiquitin carboxyl-terminal hydrolase L5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse