Anti-ACKR2 Rabbit Polyclonal Antibody
Catalog # ABGEAP2012B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Atypical Chemokine Receptor 2
- Antigen symbol:ACKR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal ( 313–328 aa)
- Cross adsorption:No
- Antigen synonyms:CCBP2|CCR10|hD6|CMKBR9|CCR9|D6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ACKR2 (atypical chemokine receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse