Anti-MLLT10 (AF10) Rabbit Polyclonal Antibody
Catalog # ABGEAP1906B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Myeloid/lymphoid or Mixed-lineage Leukemia; translocated to, 10
- Antigen symbol:MLLT10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Cross adsorption:No
- Antigen synonyms:MGC75086|DKFZp686E10210|myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog)|myeloid/lymphoid or mixed-lineage leukemia translocated to|AF10|MLLT10|translocated to|myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog|Drosophila)|10|type I AF10 protein|ALL-1 fused gene from chromosome 10
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MLLT10 (AF10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human