Anti-PDLIM1 Rabbit Polyclonal Antibody
ABGEAP14914B
: Abgent
- Antibody type:Primary
- Antigen name:PDZ And LIM Domain 1
- Antigen symbol:PDLIM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:9124
- Antigen synonyms:CLP-36|hCLIM1|CLIM1|CLP36|HEL-S-112
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:36.072 kDa
- Sequence:GVFVKLRDRHRHPEC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PDLIM1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 254-282 amino acids from the C-terminal region of human PDLIM1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: PDLIM1 (PDZ and LIM domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human