Order Entry
Ireland
ContactUsLinkComponent
Anti-KDM5B Rabbit Polyclonal Antibody
Anti-KDM5B Rabbit Polyclonal Antibody
Catalog # ABGEAP14476A
Supplier:  Abgent
Anti-KDM5B Rabbit Polyclonal Antibody
Catalog # ABGEAP14476A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lysine (K)-Specific Demethylase 5B
  • Antigen symbol:
    KDM5B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Gene ID:
    10765
  • Antigen synonyms:
    PLU1|JARID1B|CT31|RBP2-H1|PUT1|RBBP2H1A|PPP1R98|PLU-1
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    175.658 kDa
  • Sequence:
    EYKPHDIPQRQSVQPSETC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This JARID1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 192-224 amino acids from the N-terminal region of human JARID1B.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: KDM5B (lysine (K)-specific demethylase 5B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse