Order Entry
Ireland
ContactUsLinkComponent
 
Anti-PDK3 Rabbit Polyclonal Antibody
  ABGEAP14274B
 :  Abgent
Anti-PDK3 Rabbit Polyclonal Antibody
  ABGEAP14274B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Pyruvate Dehydrogenase Kinase 3
  • Antigen symbol:
    PDK3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Gene ID:
    236900
  • Antigen synonyms:
    CMTX6|pyruvate dehydrogenase kinase 3|GS1-358P8.4
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    47.923 kDa
  • Sequence:
    YKAKQDKIKSNRTC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This Mouse Pdk3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 387-414 amino acids from the C-terminal region of mouse Pdk3.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: PDK3 (pyruvate dehydrogenase kinase, isozyme 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse