Order Entry
Ireland
ContactUsLinkComponent
Anti-MAP3K3 Rabbit Polyclonal Antibody
Anti-MAP3K3 Rabbit Polyclonal Antibody
Catalog # ABGEAP14251C
Supplier:  Abgent
Anti-MAP3K3 Rabbit Polyclonal Antibody
Catalog # ABGEAP14251C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase Kinase Kinase 3
  • Antigen symbol:
    MAP3K3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Center
  • Gene ID:
    26406
  • Antigen synonyms:
    AW548911|Mekk3|MAPKKK3|mKIAA4031
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    70.776 kDa
  • Sequence:
    SQMSRARSFPDNRKEC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,47 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This Mouse Map3k3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 228-257 amino acids from the Central region of mouse Map3k3.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MAP3K3 (mitogen-activated protein kinase kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse