Anti-SLC41A2 Rabbit Polyclonal Antibody
Catalog # ABGEAP13832A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 41 Member A2
- Antigen symbol:Solute carrier family 41, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:SLC41A1-L1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SLC41A2 (solute carrier family 41 (magnesium transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human