Anti-NEUROD6 Rabbit Polyclonal Antibody
Catalog # ABGEAP13481A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Neuronal differentiation 6
- Antigen symbol:NEUROD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:Math-2|NEX1M|bHLHa2|Nex1|MATH2|Atoh2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: NEUROD6 (neuronal differentiation 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human