Anti-HSD11B1L Rabbit Polyclonal Antibody [clone: 51CT78.40.5]
Catalog # ABGEAP11478B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid 11-beta Dehydrogenase 1 Like
- Antigen symbol:HSD11B1L
- Clonality:Polyclonal
- Clone:51CT78.40.5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:PIG61|HER1|epidermal growth f|mENA|proto-oncogene c-ErbB-1|ERBB|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: HSD11B1L (hydroxysteroid (11-beta) dehydrogenase 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human