Anti-ZNF829 Rabbit Polyclonal Antibody [clone: 235CT7.8.2.3]
Catalog # ABGEAP11427C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 829
- Antigen symbol:ZNF829
- Clonality:Polyclonal
- Clone:235CT7.8.2.3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:hGWL|GWL|MAST-L|THC2|GW|GREATWALL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZNF829 (zinc finger protein 829)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human