Anti-CCDC84 Rabbit Polyclonal Antibody [clone: 206CT5.4.2]
Catalog # ABGEAP11365A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-activated Catalytic Subunit Alpha 2
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Clone:206CT5.4.2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:AMPKa2|AMPK|AMPK2|PRKAA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CCDC84 (coiled-coil domain containing 84)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse