Anti-NPS3A Rabbit Polyclonal Antibody
ABGEAP11156C
: Abgent
- Antibody type:Primary
- Antigen name:Nipsnap Homolog 3A
- Antigen symbol:NIPSNAP3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal F279 (250–350 aa)
- Cross adsorption:No
- Antigen synonyms:HSPC299|Protein NipSnap homolog 4|Protein NipSnap homolog 3A|NIPSNAP3A Antibody|TASSC|NipSnap3A|NIPSNAP4
- Sequence:F279
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: NPS3A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse