Anti-BCO2 Rabbit Polyclonal Antibody
Catalog # ABGEAP11050A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Beta-Carotene Oxygenase 2
- Antigen symbol:BCO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-Terminal (1–100 aa)
- Cross adsorption:No
- Antigen synonyms:B-DIOX-II|BCDO2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BCO2 (beta-carotene oxygenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse