Anti-SPDYE3 Rabbit Polyclonal Antibody
Catalog # ABGEAP10357C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Speedy/RINGO Cell Cycle Regulator Family Member E3
- Antigen symbol:SPDYE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:No
- Antigen synonyms:RAD9 homolog|RAD 9 (S pombe) homolog|RAD9A|Cell cycle checkpoint control protein|RAD9 homolog A
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SPDYE3 (speedy/RINGO cell cycle regulator family member E3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human