Anti-FSHB Mouse Monoclonal Antibody
Catalog # ABGEAM1973B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Follicle Stimulating Hormone Beta Subunit
- Antigen symbol:FSHB
- Clonality:Monoclonal
- Clone:365CT24.1.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Cross adsorption:No
- Gene ID:2488
- Antigen synonyms:HH24
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:14.7 kDa
- Sequence:YTRDLVYKDPARPKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This FSHB/FSH antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 36-64 amino acids from the Central region of human FSHB/FSH.
- Purification:Protein G Purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: FSHB (follicle stimulating hormone, beta polypeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Mouse
Isotype: none
Reactivity: Human