Anti-C3orf72 Rabbit Polyclonal Antibody
Catalog # AVIVARP69888_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box L2 neighbor
- Antigen symbol:FOXL2NB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:401089
- Antigen synonyms:C3orf72
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: C3orf72 (chromosome 3 open reading frame 72)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human