Anti-HLA-E Rabbit Polyclonal Antibody
Catalog # AVIVARP62847_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:major histocompatibility complex, class I, E
- Antigen symbol:HLA-E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3133
- Antigen synonyms:EA2.1|HLA-6.2|EA1.2|MHC|QA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HLA-E (major histocompatibility complex, class I, E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human