Anti-HARS2 Rabbit Polyclonal Antibody
AVIVARP62671_P050
- Antibody type:Primary
- Antigen name:histidyl-tRNA synthetase 2, mitochondrial
- Antigen symbol:HARS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23438
- Antigen synonyms:HO3|HARSR|PRLTS2|HARSL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HARS2 (histidyl-tRNA synthetase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human