Anti-OR2D3 Rabbit Polyclonal Antibody
Catalog # AVIVARP59720_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:olfactory receptor, family 2, subfamily D, member 3
- Antigen symbol:OR2D3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:120775
- Antigen synonyms:OR11-89
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OR2D3 (olfactory receptor, family 2, subfamily D, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: