Anti-OR2A5 Rabbit Polyclonal Antibody
Catalog # AVIVARP59711_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:olfactory receptor, family 2, subfamily A, member 5
- Antigen symbol:OR2A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:393046
- Antigen synonyms:OR2A26|OR7-141|OR2A8|OR2A11P|OR7-138
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OR2A5 (olfactory receptor, family 2, subfamily A, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: