Anti-ELMO1 Rabbit Polyclonal Antibody
Catalog # AVIVARP59647_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:engulfment and cell motility 1
- Antigen symbol:ELMO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9844
- Antigen synonyms:ELMO-1|CED12|CED-12
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: