Anti-ZER1 Rabbit Polyclonal Antibody
Catalog # AVIVARP59407_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zyg-11 related, cell cycle regulator
- Antigen symbol:ZER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10444
- Antigen synonyms:ZYG11BL|C9orf60|ZYG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZER1 (zyg-11 related, cell cycle regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human