Anti-CPE Rabbit Polyclonal Antibody
Catalog # AVIVARP58445_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:carboxypeptidase E
- Antigen symbol:CPE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:1363
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CPE (carboxypeptidase E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: