Anti-GRIPAP1 Rabbit Polyclonal Antibody
AVIVARP57358_P050
- Antibody type:Primary
- Antigen name:GRIP1 associated protein 1
- Antigen symbol:GRIPAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:56850
- Antigen synonyms:GRASP-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GRIPAP1 (GRIP1 associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: