Anti-ATP6V1B1 Rabbit Polyclonal Antibody
Catalog # AVIVARP56327_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1
- Antigen symbol:ATP6V1B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:525
- Antigen synonyms:RTA1B|ATP6B1|VATB|VMA2|VPP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ATP6V1B1 (ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: