Anti-MTRF1L Rabbit Polyclonal Antibody
Catalog # AVIVARP56320_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:mitochondrial translational release factor 1-like
- Antigen symbol:MTRF1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:54516
- Antigen synonyms:HMRF1L|MRF1L|mtRF1a
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MTRF1L (mitochondrial translational release factor 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: