Anti-C20orf195 Rabbit Polyclonal Antibody
AVIVARP53763_P050
- Antibody type:Primary
- Antigen name:Chromosome 20 open reading frame 195
- Antigen symbol:C20orf195
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Guinea pig
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:79025
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: C20orf195 (chromosome 20 open reading frame 195)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Guinea pig