Order Entry
Ireland
ContactUsLinkComponent
Anti-PDXK Rabbit Polyclonal Antibody
Anti-PDXK Rabbit Polyclonal Antibody
Catalog # AVIVARP53615_P050
undefined
Anti-PDXK Rabbit Polyclonal Antibody
Catalog # AVIVARP53615_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    pyridoxal (pyridoxine, vitamin B6) kinase
  • Antigen symbol:
    PDXK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    8566
  • Antigen synonyms:
    C21orf124|HEL-S-1a|PRED79|C21orf97|PNK|PKH
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: PDXK (pyridoxal (pyridoxine, vitamin B6) kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: