Anti-ALG2 Rabbit Polyclonal Antibody
AVIVARP48974_P050
- Antibody type:Primary
- Antigen name:ALG2, alpha-1,3/1,6-mannosyltransferase
- Antigen symbol:ALG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:85365
- Antigen synonyms:CDGIi|CMS14|hALPG2|CMSTA3|NET38
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ALG2 (ALG2, alpha-1,3/1,6-mannosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: