Anti-TPD52 Rabbit Polyclonal Antibody
Catalog # AVIVARP48271_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Tumor Protein D52
- Antigen symbol:TPD52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:7163
- Antigen synonyms:N8L|hD52|PC-1|D52|PrLZ
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TPD52 (tumor protein D52)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: