Anti-TPM2 Rabbit Polyclonal Antibody
Catalog # AVIVARP48224_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tropomyosin 2 (beta)
- Antigen symbol:TPM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:7169
- Antigen synonyms:DA1|HEL-S-273|DA2B|TMSB|AMCD1|NEM4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TPM2 (tropomyosin 2 (beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: