Anti-COQ2 Rabbit Polyclonal Antibody
AVIVARP47180_P050
- Antibody type:Primary
- Antigen name:coenzyme Q2 4-hydroxybenzoate polyprenyltransferase
- Antigen symbol:COQ2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:27235
- Antigen synonyms:COQ10D1|PHB:PPT|CL640|MSA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: COQ2 (coenzyme Q2 4-hydroxybenzoate polyprenyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: