Anti-ATP1B4 Rabbit Polyclonal Antibody
Catalog # AVIVARP46848_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ATPase, Na+/K+ transporting, beta 4 polypeptide
- Antigen symbol:ATP1B4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:23439
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ATP1B4 (ATPase, Na+/K+ transporting, beta 4 polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: