Order Entry
Ireland
ContactUsLinkComponent
Anti-RTN4 Rabbit Polyclonal Antibody
Anti-RTN4 Rabbit Polyclonal Antibody
  AVIVARP46812_P050
Anti-RTN4 Rabbit Polyclonal Antibody
  AVIVARP46812_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    reticulon 4
  • Antigen symbol:
    RTN4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    57142
  • Antigen synonyms:
    ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: RTN4 (reticulon 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: