Order Entry
Ireland
ContactUsLinkComponent
Anti-ATP6V0A1 Rabbit Polyclonal Antibody
Anti-ATP6V0A1 Rabbit Polyclonal Antibody
Catalog # AVIVARP46581_P050
Anti-ATP6V0A1 Rabbit Polyclonal Antibody
Catalog # AVIVARP46581_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATPase, H+ transporting, lysosomal V0 subunit a1
  • Antigen symbol:
    ATP6V0A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    535
  • Antigen synonyms:
    VPP1|a1|Stv1|ATP6N1A|Vph1|ATP6N1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: ATP6V0A1 (ATPase, H+ transporting, lysosomal V0 subunit a1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: