Anti-NUSAP1 Rabbit Polyclonal Antibody
AVIVARP46285_P050
- Antibody type:Primary
- Antigen name:nucleolar and spindle associated protein 1
- Antigen symbol:NUSAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Pig,Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:51203
- Antigen synonyms:LNP|Q0310|ANKT|NUSAP|BM037|SAPL|PRO0310p1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NUSAP1 (nucleolar and spindle associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Pig