Anti-DESI1 Rabbit Polyclonal Antibody
Catalog # AVIVARP46230_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:desumoylating isopeptidase 1
- Antigen symbol:DESI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:27351
- Antigen synonyms:D15Wsu75e|DESI2|DeSI-1|DJ347H13.4|FAM152B|PPPDE2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: DESI1 (desumoylating isopeptidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human