Anti-PRDM15 Rabbit Polyclonal Antibody
AVIVARP45785_P050
- Antibody type:Primary
- Antigen name:PR domain containing 15
- Antigen symbol:PRDM15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:63977
- Antigen synonyms:C21orf83|ZNF298|PFM15
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PRDM15 (PR domain containing 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat