Anti-PTPRR Rabbit Polyclonal Antibody
AVIVARP45386_P050
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type R
- Antigen symbol:PTPRR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5801
- Antigen synonyms:PTP-SL|EC-PTP|PCPTP1|PTPRQ|PTPBR7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PTPRR (protein tyrosine phosphatase, receptor type, R)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: