Anti-ACVR2B Rabbit Polyclonal Antibody
Catalog # AVIVARP45043_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:activin A receptor, type IIB
- Antigen symbol:ACVR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:93
- Antigen synonyms:ActR-IIB|ACTRIIB|HTX4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ACVR2B (activin A receptor, type IIB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: