Anti-D4S234E Rabbit Polyclonal Antibody
Catalog # AVIVARP44912_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:neuron specific gene family member 1
- Antigen symbol:NSG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:27065
- Antigen synonyms:P21|D4S234|D4S234E|NEEP21
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: D4S234E
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human